Proteintech
Manufacturer Code:551131AP150UL
Catalog # 50-173-5992
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMDM ANP B ANPR B ANPRB GC B Guanylate cyclase B GUC2B GUCY2B Natriuretic Peptide Receptor B NPR2 NPRB; ANP-B; ANPR-B; atrial natriuretic peptide B-type receptor; Atrial natriuretic peptide receptor 2; Atrial natriuretic peptide receptor type B; GC-B; guanylate cyclase 2B; Guanylate cyclase B; guanylyl cyclase-B; natriuretic peptide receptor B type; natriuretic peptide receptor B/guanylate cyclase B (atrionatriuretic peptide receptor B); Nppb receptor; NPR-B
Gene Aliases: AMDM; ANPb; ANPRB; cn; ECDM; GC-B; GC-B2; GC-B3; GUC2B; GUCY2B; mNPR-B; NPR2; NPRB; NPRBi; pwe; SNSK
UniProt ID: (Human) P20594, (Mouse) Q6VVW5
Entrez Gene ID: (Human) 4882, (Mouse) 230103
Molecular Function: adenylate cyclase cyclase guanylate cyclase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.