OriGene
Manufacturer Code:CF809079
Catalog # 50-166-9252
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI11C3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 27-102 of human NPPB produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BNP; BNP(1-28); BNP(1-29); BNP(1-30); BNP(1-32); BNP(2-31); BNP(3-29); BNP(3-30); BNP(3-32); BNP(4-27); BNP(4-29); BNP(4-30); BNP(4-31); BNP(4-32); BNP(5-29); BNP(5-31); BNP(5-32); Brain natriuretic factor prohormone; Brain natriuretic peptide; Brain natriuretic peptide 29; Brain natriuretic peptide 32; brain type natriuretic peptide; des-SerPro-BNP; Gamma-brain natriuretic peptide; Iso-ANP; natriuretic peptide precursor B; Natriuretic peptides B; natriuretic protein; NT-pro-BNP; NT-proBNP; NT-proBNP(1-76); preproBNP; proBNP(3-108); proBNP(79-108)
Gene Aliases: BNP; NPPB
UniProt ID: (Human) P16860
Entrez Gene ID: (Human) 4879
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.