Abnova
Manufacturer Code:PAB5082
Catalog # 89-131-729
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to rat Nppb. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Nppb.
For Research Use Only
Protein Aliases: 5 kDa cardiac natriuretic peptide; Bnf; BNP; BNP-45; Brain natriuretic factor; Brain natriuretic factor prohormone; Brain natriuretic peptide; Brain natriuretic peptide 45; Gamma-brain natriuretic peptide; Iso-ANP; natriuretic peptide precursor B; natriuretic peptide precursor type B; Natriuretic peptides B; preproBNP
Gene Aliases: Bnf; BNP; Nppb
UniProt ID: (Rat) P13205
Entrez Gene ID: (Rat) 25105
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.