Invitrogen
Manufacturer Code:PA547610
Catalog # PIPA547610
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Nephronectin Asp20-Cys561 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EGFL6L Nctn NPNT POEM nephronectin; Nephronectin; Preosteoblast EGF-like repeat protein with MAM domain
Gene Aliases: 1110009H02Rik; AA682063; AI314031; Nctn; Neph1; Npnt; POEM
UniProt ID: (Mouse) Q91V88
Entrez Gene ID: (Mouse) 114249
Molecular Function: extracellular matrix protein extracellular matrix structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.