Invitrogen
Manufacturer Code:PA5100647
Catalog # PIPA5100647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NPLOC4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AK129375 FLJ20657 homolog of yeast nuclear protein localization 4 KIAA1499 mKIAA1499 Npl4 NPL4 homolog ubiquitin recognition factor NPLOC4 NPLOC4 ubiquitin recognition factor nuclear protein localization 4 homolog nuclear protein localization 4 homolog (S. cerevisiae) nuclear protein localization protein 4 homolog Protein NPL4; homolog of yeast nuclear protein localization 4; NPLOC4 ubiquitin recognition factor; nuclear protein localization 4 homolog; Nuclear protein localization protein 4 homolog; Protein NPL4
Gene Aliases: AK129375; KIAA1499; mKIAA1499; NPL4; NPLOC4
UniProt ID: (Human) Q8TAT6, (Mouse) P60670, (Rat) Q9ES54
Entrez Gene ID: (Human) 55666, (Mouse) 217365, (Rat) 140639
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.