Invitrogen
Manufacturer Code:PA5113402
Catalog # PIPA5113402
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human N-acetylneuraminate lyase protein (17-77AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610033B02Rik C112 C1orf13 dihydrodipicolinate synthetase homolog 1 N-acetylneuraminate lyase N-acetylneuraminate pyruvate lyase N-acetylneuraminate pyruvate lyase (dihydrodipicolinate synthase) N-acetylneuraminate pyruvate-lyase N-acetylneuraminic acid aldolase NAL NALase Npl NPL1 Sialate lyase Sialate-pyruvate lyase Sialic acid aldolase Sialic acid lyase; dihydrodipicolinate synthetase homolog 1; N-acetylneuraminate lyase; N-acetylneuraminate pyruvate lyase (dihydrodipicolinate synthase); N-acetylneuraminate pyruvate-lyase; N-acetylneuraminic acid aldolase; NALase; Sialate lyase; Sialate-pyruvate lyase; Sialic acid aldolase; Sialic acid lyase
Gene Aliases: 0610033B02Rik; C112; C1orf13; NAL; NPL; NPL1
UniProt ID: (Human) Q9BXD5, (Mouse) Q9DCJ9
Entrez Gene ID: (Human) 80896, (Mouse) 74091
Molecular Function: lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.