Novus Biologicals
Manufacturer Code:NBP29311220UL
Catalog # NB028114
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human NPL (NP_110396.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C112 C1orf13MGC61869 chromosome 1 open reading frame 13 DHDPS1 dihydrodipicolinate synthetase homolog 1 EC 4.1.3.3 MGC149582 N-acetylneuraminate lyase N-acetylneuraminate pyruvate lyase (dihydrodipicolinate synthase) N-acetylneuraminate pyruvate-lyase N-acetylneuraminic acid aldolase NAL NALase NPL1 Sialate lyase Sialate-pyruvate lyase Sialic acid aldolase Sialic acid lyase; dihydrodipicolinate synthetase homolog 1; N-acetylneuraminate lyase; N-acetylneuraminate pyruvate lyase (dihydrodipicolinate synthase); N-acetylneuraminate pyruvate-lyase; N-acetylneuraminic acid aldolase; NALase; Sialate lyase; Sialate-pyruvate lyase; Sialic acid aldolase; Sialic acid lyase
Gene Aliases: C112; C1orf13; NAL; NPL; NPL1
UniProt ID: (Human) Q9BXD5
Entrez Gene ID: (Human) 80896
Molecular Function: lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.