Novus Biologicals
Manufacturer Code:NBP247626
Catalog # NBP247626
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide from human NPHP4 (aa877-926). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ46306 KIAA0673nephrocystin-4 nephrocystin 4 nephronophthisis 4 Nephroretinin POC10 POC10 centriolar protein homolog SLSN4nephroretinin; Nephrocystin-4; Nephroretinin; POC10 centriolar protein homolog
Gene Aliases: KIAA0673; NPHP4; POC10; SLSN4
UniProt ID: (Human) O75161
Entrez Gene ID: (Human) 261734
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.