Invitrogen
Manufacturer Code:MA524451
Catalog # PIMA524451
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3B1 |
Immunogen | NPHP3 (NP&lowbar694972.3 106 a.a. &bsim 205 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C,Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cilia and flagella associated protein 31; Meckel syndrome, type 7; Nephrocystin-3; Nephrocystin-3 NPHP3 KIAA2000
Gene Aliases: CFAP31; KIAA2000; MKS7; NPH3; NPHP3; RHPD; RHPD1; SLSN3
UniProt ID: (Human) Q7Z494
Entrez Gene ID: (Human) 27031
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.