Invitrogen
Manufacturer Code:PA5113061
Catalog # PIPA5113061
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Nephrocystin-3 protein (1-130AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3632410F03Rik AI550417 C230078J01 CFAP31 cilia and flagella associated protein 31 D330020E01Rik KIAA2000 Meckel syndrome type 7 MKS7 Nephrocystin-3 nephronophthisis 3 (adolescent) NPH3 Nphp3 pcy RHPD RHPD1 SLSN3; cilia and flagella associated protein 31; Meckel syndrome, type 7; Nephrocystin-3
Gene Aliases: 3632410F03Rik; AI550417; C230078J01; CFAP31; D330020E01Rik; KIAA2000; MKS7; NPH3; NPHP3; pcy; RHPD; RHPD1; SLSN3
UniProt ID: (Human) Q7Z494, (Mouse) Q7TNH6
Entrez Gene ID: (Human) 27031, (Mouse) 74025
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.