Novus Biologicals
Manufacturer Code:NBP183543
Catalog # NBP183543
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:PEFAHSSIDVEGPFANVNRDDWDIAVASLLQVTPLFSHSLWSNTVRCYLIYTDETQPEMDLFLKDYSPKLKRMCETMGYFFHAVYFPIDVENQYLTVRKWEIEKS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cilia and flagella associated protein 31; FLJ30691 FLJ36696 KIAA2000DKFZp667K242 MKS7DKFZp781K1312 nephrocystin-3 nephronophthisis 3 (adolescent) NPH3MGC78666 RHPD; Meckel syndrome, type 7; Nephrocystin-3
Gene Aliases: CFAP31; KIAA2000; MKS7; NPH3; NPHP3; RHPD; RHPD1; SLSN3
UniProt ID: (Human) Q7Z494
Entrez Gene ID: (Human) 27031
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.