Invitrogen
Manufacturer Code:PA537187
Catalog # PIPA537187
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues in human Nephrocystin-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Juvenile nephronophthisis 1 protein; nephrocystin 1; Nephrocystin-1; Nephrocystin1 Nephrocystin 1 Juvenile nephronophthisis 1 protein NPHP1 NPH1; nephronophthisis 1 (juvenile); nephronophthisis 1 (juvenile) homolog
Gene Aliases: JBTS4; NPH1; NPHP1; SLSN1
UniProt ID: (Human) O15259, (Mouse) Q9QY53
Entrez Gene ID: (Human) 4867, (Rat) 296136, (Mouse) 53885
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.