Proteintech
Manufacturer Code:195051AP150UL
Catalog # 50-173-2422
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein coupled receptor 74 GPR74 HLWAR77 neuropeptide FF receptor 2 NPFFR2; G protein-coupled receptor 74; G-protein coupled receptor 74; G-protein coupled receptor HLWAR77; neuropeptide FF 2; Neuropeptide FF receptor 2; Neuropeptide G-protein coupled receptor
Gene Aliases: GPR74; HLWAR77; NPFF2; NPFFR2; NPGPR
UniProt ID: (Human) Q9Y5X5
Entrez Gene ID: (Human) 10886
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.