Invitrogen
Manufacturer Code:PA5111081
Catalog # PIPA5111081
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human NPFF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ10751 GPR147 G-protein coupled receptor 147 Neuropeptide FF receptor 1 NPFF1 NPFF1 receptor NPFF1R1 NPFFR1 NPFF-R1 OT7T022 RFamide-related peptide receptor OT7T022; G protein-coupled receptor 147; G-protein coupled receptor 147; neuropeptide FF 1; Neuropeptide FF receptor 1; RFamide-related peptide receptor OT7T022
Gene Aliases: GPR147; NPFF1; NPFF1R1; NPFFR1; OT7T022
UniProt ID: (Human) Q9GZQ6
Entrez Gene ID: (Human) 64106
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.