Invitrogen
Manufacturer Code:PA5115244
Catalog # PIPA5115244
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human FMRFamide corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FMRFAL FMRFamide-related peptides Neuropeptide AF Neuropeptide AF-like Neuropeptide FF neuropeptide FF-amide peptide precursor Neuropeptide SF NPAF Npff NPSF Pro-FMRFamide-related neuropeptide FF; FMRFamide-related peptides; Neuropeptide AF; Neuropeptide FF; Neuropeptide SF; NPAF; NPFF; NPSF; Pro-FMRFamide-related neuropeptide FF
Gene Aliases: FMRFAL; NPFF
UniProt ID: (Human) O15130
Entrez Gene ID: (Human) 8620
Molecular Function: neuropeptide peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.