Proteintech
Manufacturer Code:172111AP150UL
Catalog # 50-173-1443
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NPEPL1 Fusion Protein Ag10994 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aminopeptidase like 1 KIAA1974 NPEPL1 Probable aminopeptidase NPEPL1; Aminopeptidase-like 1; Probable aminopeptidase NPEPL1
Gene Aliases: bA261P9.2; BC023239; C85514; KIAA1974; mKIAA1974; NPEPL1
UniProt ID: (Human) Q8NDH3, (Mouse) Q6NSR8
Entrez Gene ID: (Human) 79716, (Mouse) 228961, (Rat) 311671
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.