Invitrogen
Manufacturer Code:PA542982
Catalog # PIPA542982
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human NPDC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neural proliferation differentiation and control 1 CAB CAB- CAB-1 CAB1 DKFZP586J0523 NPDC-1; Neural proliferation differentiation and control protein 1; neural proliferation differentiation control-1 protein; neural proliferation, differentiation and control gene 1; neural proliferation, differentiation and control, 1; NPDC-1
Gene Aliases: AI314472; CAB; CAB-; CAB-1; CAB1; NPDC-1; NPDC1
UniProt ID: (Human) Q9NQX5, (Mouse) Q64322
Entrez Gene ID: (Bovine) 100125919, (Human) 56654, (Dog) 480668, (Mouse) 18146, (Rat) 296562
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.