Abnova
Manufacturer Code:H00010577M01
Catalog # 89-002-772
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4B9 |
Immunogen | NPC2 (AAH02532 1 a.a. ~ 151 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant NPC2.
For Research Use Only
Protein Aliases: epididymal protein 1; Epididymal secretory protein E1; HE1; Human epididymis-specific protein 1; MGC1333; Niemann-Pick disease type C2 protein; Niemann-Pick disease, type C2; NP-C2; NPC intracellular cholesterol transporter 2; tissue-specific secretory protein
Gene Aliases: EDDM1; HE1; NPC2
UniProt ID: (Human) P61916
Entrez Gene ID: (Human) 10577
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.