Invitrogen
Manufacturer Code:PA581613
Catalog # PIPA581613
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Rat NPC2 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymal secretory protein 1; epididymal secretory protein E1; Niemann Pick type C2; NPC2 Niemann-Pick disease type C2 EDDM1 HE1 MGC1333 NP-C2 epididymal protein 1 epididymal secretory protein epididymal secretory protein E1 tissue-specific secretory protein
Gene Aliases: re1
Entrez Gene ID: (Rat) 286898
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.