Invitrogen
Manufacturer Code:PA551463
Catalog # PIPA551463
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NPC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymal protein 1; epididymal secretory protein 1; Epididymal secretory protein E1; He1; Human epididymis-specific protein 1; mE1; Niemann Pick type C2; Niemann Pick type C2 protein homolog; Niemann-Pick disease type C2 protein; Niemann-Pick disease, type C2; NPC intracellular cholesterol transporter 2; NPC2 Niemann-Pick disease type C2 EDDM1 HE1 MGC1333 NP-C2 epididymal protein 1 epididymal secretory protein epididymal secretory protein E1 tissue-specific secretory protein; tissue-specific secretory protein
Gene Aliases: 2700012J19Rik; AA408070; AU045843; EDDM1; HE1; NPC2; re1
UniProt ID: (Human) P61916, (Mouse) Q9Z0J0
Entrez Gene ID: (Human) 10577, (Mouse) 67963, (Rat) 286898
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.