Abnova
Manufacturer Code:PAB19731
Catalog # 89-121-259
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 21-33 at internal region of mouse Npc2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of Npc2.
For Research Use Only
Protein Aliases: 2700012J19Rik; AA408070; AU045843; Epididymal secretory protein E1; HE1; mE1; Niemann Pick type C2 protein homolog; NPC intracellular cholesterol transporter 2
Gene Aliases: 2700012J19Rik; AA408070; AU045843; HE1; Npc2
UniProt ID: (Mouse) Q9Z0J0
Entrez Gene ID: (Mouse) 67963
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.