Proteintech
Manufacturer Code:198881AP150UL
Catalog # 50-173-2585
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NPC2 Fusion Protein Ag13719 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymal protein 1; Epididymal secretory protein E1; He1; HE1 Niemann Pick disease type C2 NPC2; Human epididymis-specific protein 1; mE1; Niemann Pick type C2 protein homolog; Niemann-Pick disease type C2 protein; Niemann-Pick disease, type C2; NPC intracellular cholesterol transporter 2; tissue-specific secretory protein
Gene Aliases: 2700012J19Rik; AA408070; AU045843; EDDM1; HE1; NPC2
UniProt ID: (Human) P61916, (Mouse) Q9Z0J0
Entrez Gene ID: (Human) 10577, (Mouse) 67963
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.