Invitrogen
Manufacturer Code:PA116800
Catalog # APPA6800
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunomicroscopy (IM) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal region of rat NPC1L1(between residues 1000-1100). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Niemann-Pick C1-like protein 1; Niemann-Pick disease, type C1; Niemann-Pick disease, type C1-like 1; NPC1 (Niemann-Pick disease, type C1, gene)-like 1; NPC1 like 1; NPC1-like 1; NPC1-like intracellular cholesterol transporter 1; NPC3 Niemann-Pick C3 NPC1L1 Niemann-Pick type C1 Like 1 Protein BC 400-127
Gene Aliases: 9130221N23Rik; Gm243; NPC11L1; NPC1L1
UniProt ID: (Human) Q9UHC9, (Rat) Q6T3U3
Entrez Gene ID: (Human) 29881, (Pig) 100515982, (Mouse) 237636, (Rat) 432367
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.