Invitrogen
Manufacturer Code:PA116817
Catalog # APPA6817
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunomicroscopy (IM) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to the C-terminal region of human NPC1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ98532 NPC; Niemann Pick type C1; Niemann-Pick C disease protein; Niemann-Pick C1 protein; Niemann-Pick disease, type C1; Niemann-Pick type C1; Niemann-Pick type C1 protein; NPC intracellular cholesterol transporter 1; sphingomyelinosis; truncated Niemann-Pick C1
Gene Aliases: A430089E03Rik; C85354; Cdig2; D18Ertd139e; D18Ertd723e; I79_014078; lcsd; nmf164; NPC; NPC1; spm
UniProt ID: (Pig) P56941, (Human) O15118, (Mouse) O35604
Entrez Gene ID: (Pig) 397591, (Human) 4864, (Mouse) 18145, (Chinese hamster) 100689424, (Rat) 266732
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.