Proteintech
Manufacturer Code:246251AP150UL
Catalog # 50-173-4461
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NPBWR1 Fusion Protein Ag16604 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein coupled receptor 7 GPR7 neuropeptides B/W receptor 1 NPBWR1; G protein-coupled receptor 7; G-protein coupled receptor 7; neuropeptide B/W receptor 1; Neuropeptides B/W receptor type 1; opioid-somatostatin-like receptor 7
Gene Aliases: GPR7; NPBWR1
UniProt ID: (Human) P48145
Entrez Gene ID: (Human) 2831
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.