Novus Biologicals
Manufacturer Code:NBP188622
Catalog # NBP188622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SQVELTGSSVFDYVHPGDHVEMAEQLGMKLPPGRGLLSQGTAEDGASSASSSSQSETPEPVESTSPSLLTTDNTLERSFFIRMKSTLTKRGVHIKSSGYKV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basic-helix-loop-helix-PAS protein MOP6; Basic-helix-loop-helix-PAS protein MOP6 BHLHE12 bHLHe12FLJ11605 Class E basic helix-loop-helix protein 12 Member of PAS protein 6 MOP6FLJ10003 neuronal PAS domain protein 3 neuronal PAS domain-containing protein 3 Neuronal PAS3 PAS domain-containing protein 6 PASD6FLJ11138; bHLHe12; Class E basic helix-loop-helix protein 12; Member of PAS protein 6; Neuronal PAS domain-containing protein 3; Neuronal PAS3; PAS domain-containing protein 6
Gene Aliases: BHLHE12; MOP6; NPAS3; PASD6
UniProt ID: (Human) Q8IXF0
Entrez Gene ID: (Human) 64067
Molecular Function: basic helix-loop-helix transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.