Invitrogen
Manufacturer Code:PA554053
Catalog # PIPA554053
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NPAS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basic-helix-loop-helix-PAS protein MOP4; bHLHe9; Class E basic helix-loop-helix protein 9; Member of PAS protein 4; member of PAS superfamily 4; MOP4 PASD4 bHLHe9 PAS domain-containing protein 4 basic-helix-loop-helix-PAS protein MOP4 class E basic helix-loop-helix protein 9 member of PAS protein 4 member of PAS superfamily 4 neuronal PAS domain-containing protein 2 neuronal PAS2; Neuronal PAS domain-containing protein 2; Neuronal PAS2; PAS domain-containing protein 4
Gene Aliases: BHLHE9; MOP4; NPAS2; PASD4
UniProt ID: (Human) Q99743, (Mouse) P97460
Entrez Gene ID: (Human) 4862, (Mouse) 18143
Molecular Function:
basic helix-loop-helix transcription factor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.