Invitrogen
Manufacturer Code:PA578083
Catalog # PIPA578083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human NPAS1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basic-helix-loop-helix-PAS protein MOP5; bHLHe11; bHLHe11 MOP5 PASD5 neuronal PAS domain protein 1; Class E basic helix-loop-helix protein 11; Member of PAS protein 5; member of PAS superfamily 5; Neuronal PAS domain-containing protein 1; Neuronal PAS1; PAS domain-containing protein 5
Gene Aliases: BHLHE11; MOP5; NPAS1; PASD5
UniProt ID: (Human) Q99742, (Mouse) P97459
Entrez Gene ID: (Human) 4861, (Mouse) 18142, (Rat) 308387
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.