Novus Biologicals
Manufacturer Code:NBP243651
Catalog # NBP243651
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human NPAS1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basic-helix-loop-helix-PAS protein MOP5; Basic-helix-loop-helix-PAS protein MOP5 BHLHE11 bHLHe11member of PAS superfamily 5 Class E basic helix-loop-helix protein 11 Member of PAS protein 5 MOP5Neuronal PAS1 neuronal PAS domain protein 1 neuronal PAS domain-containing protein 1 PASD5PAS domain-containing protein 5; bHLHe11; Class E basic helix-loop-helix protein 11; Member of PAS protein 5; member of PAS superfamily 5; Neuronal PAS domain-containing protein 1; Neuronal PAS1; PAS domain-containing protein 5
Gene Aliases: BHLHE11; MOP5; NPAS1; PASD5
UniProt ID: (Human) Q99742
Entrez Gene ID: (Human) 4861
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.