Novus Biologicals
Manufacturer Code:NBP293567100UL
Catalog # NB029007
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 121-370 of human NOXO1 (NP_653204.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC20258 NADPH oxidase organizer 1 Nox organizer 1 Nox-organizing protein 1 P41NOX P41NOXA P41NOXB P41NOXC regulatory protein P41NOX SH3 and PX domain-containing protein 5 SH3PXD5NADPH oxidase regulatory protein SNX28; NADPH oxidase organizer 1; NADPH oxidase regulatory protein; Nox organizer 1; Nox-organizing protein 1; regulatory protein P41NOX; SH3 and PX domain-containing protein 5
Gene Aliases: NOXO1; P41NOX; P41NOXA; P41NOXB; P41NOXC; SH3PXD5; SNX28
UniProt ID: (Human) Q8NFA2
Entrez Gene ID: (Human) 124056
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.