Proteintech
Manufacturer Code:155511AP150UL
Catalog # 50-173-0309
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NOXA2/p67phox Fusion Protein Ag7909 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 67 kDa neutrophil oxidase factor; chronic granulomatous disease, autosomal 2; NADPH oxidase activator 2; NADPH oxidase activator 2 NCF 2 NCF2 Neutrophil cytosol factor 2 neutrophil cytosolic factor 2 NOXA2 P67 PHOX p67phox; NADPH oxidase subunit (67 kD); NADPH oxidase subunit (67kDa); NCF-2; ncf2; ncf2 {ECO:0000312|EMBL:FAA00361.1}; Neutrophil cytosol factor 2; neutrophil cytosolic factor 2 (65kD, chronic granulomatous disease, autosomal 2); Neutrophil NADPH oxidase factor 2; p67-phox
Gene Aliases: NCF-2; NCF2; NOXA2; P67-PHOX; P67PHOX
UniProt ID: (Human) P19878, (Mouse) O70145, (Rat) A7E3N2
Entrez Gene ID: (Human) 4688, (Mouse) 17970, (Rat) 364018
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.