Invitrogen
Manufacturer Code:PA595084
Catalog # PIPA595084
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human NOXA1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antigen NY-CO-31; inhibitory NADPH oxidase activator 1; NADPH oxidase activator 1; NADPH oxidase activator 1 NOX activator 1 Antigen NY-CO-31 NCF2-like protein P67phox-like factor p51-nox NOXA1 P51NOX; NCF2-like protein; NOX activator 1; p51-nox; P67phox-like factor; serologically defined colon cancer antigen 31
Gene Aliases: NOXA1; NY-CO-31; P51NOX; SDCCAG31
UniProt ID: (Human) Q86UR1
Entrez Gene ID: (Human) 10811
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.