Invitrogen
Manufacturer Code:PA526572
Catalog # PIPA526572
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 449-478 amino acids from the C-terminal region of human NOXA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antigen NY-CO-31; inhibitory NADPH oxidase activator 1; NADPH oxidase activator 1; NCF2-like protein; NOX activator 1; NY-CO-31 SDCCAG31 p51NOX NCF2-like protein Nox activator 1 antigen NY-CO-31 inhibitory NADPH oxidase activator 1 p51-nox p67phox-like factor serologically defined colon cancer antigen 31; p51-nox; P67phox-like factor; serologically defined colon cancer antigen 31
Gene Aliases: NOXA1; NY-CO-31; P51NOX; SDCCAG31
UniProt ID: (Human) Q86UR1
Entrez Gene ID: (Human) 10811
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.