Invitrogen
Manufacturer Code:PA5112629
Catalog # PIPA5112629
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human NADPH oxidase activator 1 protein (150-313AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antigen NY-CO-31; Antigen NY-CO-31 inhibitory NADPH oxidase activator 1 NADPH oxidase activator 1 NCF2-like protein NOX activator 1 NOXA1 NY-CO-31 P51NOX p51-nox P67phox-like factor SDCCAG31 serologically defined colon cancer antigen 31; inhibitory NADPH oxidase activator 1; NADPH oxidase activator 1; NCF2-like protein; NOX activator 1; p51-nox; P67phox-like factor; serologically defined colon cancer antigen 31
Gene Aliases: NOXA1; NY-CO-31; P51NOX; SDCCAG31
UniProt ID: (Human) Q86UR1, (Mouse) Q8CJ00
Entrez Gene ID: (Human) 10811, (Mouse) 241275
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.