Novus Biologicals
Manufacturer Code:NBP293004100UL
Catalog # NB027904
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 661-765 of human NOX5 (NP_078781.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.6.3.- MGC149776 MGC149777 NADPH oxidase 5 NADPH oxidase EF-hand calcium binding domain 5 NOX5A NOX5B; NADPH oxidase 5
Gene Aliases: NOX5
UniProt ID: (Human) Q96PH1
Entrez Gene ID: (Human) 79400
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.