Abnova
Manufacturer Code:MAB14857
Catalog # 89-034-940
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | NOX4/1245 |
Immunogen | Recombinant protein corresponding to full length human NOX4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Kidney oxidase-1; Kidney superoxide-producing NADPH oxidase; KOX; KOX-1; NADPH oxidase 4; Renal NAD(P)H-oxidase; RENOX
Gene Aliases: KOX; KOX-1; NOX4; RENOX
UniProt ID: (Human) Q9NPH5
Entrez Gene ID: (Human) 50507
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.