Invitrogen
Manufacturer Code:MA535385
Catalog # PIMA535385
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0815 |
Immunogen | A synthesized peptide derived from human NOX4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AI648021 BOS_24390 Kidney oxidase-1 Kidney superoxide-producing NADPH oxidase Kox KOX1 KOX-1 NADPH oxidase 4 NOX4 Renal NAD(P)H-oxidase Renox Superoxide-generating NADPH oxidase 4; Kidney oxidase-1; Kidney superoxide-producing NADPH oxidase; KOX-1; NADPH oxidase 4; Renal NAD(P)H-oxidase; Superoxide-generating NADPH oxidase 4
Gene Aliases: AI648021; Nox4; Renox
UniProt ID: (Mouse) Q9JHI8
Entrez Gene ID: (Mouse) 50490
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.