Invitrogen
Manufacturer Code:PA576073
Catalog # PIPA576073
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human NOX4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Kidney oxidase-1; Kidney superoxide-producing NADPH oxidase; KOX-1; NADPH oxidase 4; NADPH oxidase 4 Kidney oxidase-1 KOX-1 KOX1 Kidney superoxide-producing NADPH oxidase Renal NAD(P)H-oxidase NOX4 RENOX; Renal NAD(P)H-oxidase; Superoxide-generating NADPH oxidase 4
Gene Aliases: AI648021; KOX; KOX-1; NOX4; RENOX
UniProt ID: (Human) Q9NPH5, (Mouse) Q9JHI8
Entrez Gene ID: (Human) 50507, (Mouse) 50490
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.