Invitrogen
Manufacturer Code:PA572922
Catalog # PIPA572922
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal region of the human NOX4 protein (between residues 100-200) &lsqbUniProt Q9NPH5]. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Kidney oxidase-1; Kidney superoxide-producing NADPH oxidase; KOX KOX-1 RENOX kidney oxidase-1 kidney superoxide-producing NADPH oxidase renal NAD(P)H-oxidase; KOX-1; NADPH oxidase 4; nox4; Renal NAD(P)H-oxidase; Superoxide-generating NADPH oxidase 4
Gene Aliases: AI648021; KOX; KOX-1; NOX4; RENOX
UniProt ID: (Human) Q9NPH5, (Rat) Q924V1, (Mouse) Q9JHI8
Entrez Gene ID: (Pig) 100523323, (Bovine) 378474, (Rabbit) 100339697, (Human) 50507, (Rat) 85431, (Mouse) 50490
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.