Proteintech
Manufacturer Code:200651AP150UL
Catalog # 50-173-2679
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GP91 3 gp91phox homolog 3 Mitogenic oxidase 2 MOX 2 MOX2 NADPH oxidase 3 NOX3; GP91-3; gp91phox homolog 3; head-tilt; Mitogenic oxidase 2; MOX-2; NADPH oxidase 3; NADPH oxidase catalytic subunit-like 3
Gene Aliases: GP91-3; het; MOX-2; MOX2; nmf250; NOX3
UniProt ID: (Human) Q9HBY0, (Mouse) Q672J9
Entrez Gene ID: (Human) 50508, (Mouse) 224480
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.