Proteintech
Manufacturer Code:190131AP150UL
Catalog # 50-173-2322
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NOX2 Fusion Protein Ag5536 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CGD91-phox; CYBB Cytochrome b 245 heavy chain NOX2 p91 PHOX; Cytochrome b(558) subunit beta; cytochrome b-245 beta polypeptide; Cytochrome b-245 heavy chain; cytochrome b-245, beta polypeptide; Cytochrome b558 subunit beta; endothelial type gp91-phox; gp91-1; gp91-phox; Heme-binding membrane glycoprotein gp91phox; NADPH oxidase 2; Neutrophil cytochrome b 91 kDa polypeptide; p22 phagocyte B-cytochrome; Superoxide-generating NADPH oxidase heavy chain subunit
Gene Aliases: AMCBX2; C88302; CGD; CGD91-phox; CYBB; Cyd; GP91-1; GP91-PHOX; GP91PHOX; IMD34; NOX2; p91-PHOX
UniProt ID: (Human) P04839, (Mouse) Q61093
Entrez Gene ID: (Human) 1536, (Rat) 66021, (Mouse) 13058
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.