Novus Biologicals
Manufacturer Code:NBP219568
Catalog # NBP219568
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fragment corresponding to a region within amino acids 201 and 456 of Mox1 (Uniprot ID No. Q9Y5S8) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
For Research Use Only
Protein Aliases: mitogenic oxidase (pyridine nucleotide-dependent superoxide-generating); Mitogenic oxidase 1; MOX-1; NADH/NADPH mitogenic oxidase subunit P65-MOX; NADPH oxidase 1; NADPH oxidase homolog-1; NOH-1; NOX-1
Gene Aliases: GP91-2; MOX1; NOH-1; NOH1; NOX1
UniProt ID: (Human) Q9Y5S8
Entrez Gene ID: (Human) 27035
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.