Invitrogen
Manufacturer Code:PA580859
Catalog # PIPA580859
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human CCN3/NOV/IGFBP9 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCN family member 3; CCN3 IGFBP9 CCN family member 3 IBP-9 IGF-binding protein 9 IGFBP-9 insulin-like growth factor-binding protein 9 nephroblastoma-overexpressed gene protein homolog novH protein NOV homolog; Cellular communication network factor 3; IBP-9; IGF-binding protein 9; Insulin-like growth factor-binding protein 9; Nephro blastoma-overexpressed gene protein homolog; nephroblastoma-overexpressed gene protein homolog; NovH; Protein NOV homolog
Gene Aliases: CCN3; IBP-9; IGFBP-9; IGFBP9; NOV; NOVH
UniProt ID: (Human) P48745
Entrez Gene ID: (Human) 4856
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.