Novus Biologicals
Manufacturer Code:NBP294699100UL
Catalog # NB023922
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 20-320 of human NOTUM (NP_848588.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [Wnt protein] O-palmitoleoyl-L-serine hydrolase; EC 3.- notum pectinacetylesterase homolog (Drosophila) protein notum homolog; hNOTUM; notum pectinacetylesterase homolog; Palmitoleoyl-protein carboxylesterase NOTUM; protein notum homolog
Gene Aliases: hNOTUM; NOTUM; OK/SW-CL.30
UniProt ID: (Human) Q6P988
Entrez Gene ID: (Human) 147111
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.