Invitrogen
Manufacturer Code:MA524274
Catalog # PIMA524274
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 605724 |
Immunogen | CHO-derived recombit mouse Notch-2 Leu26-Val528 (predicted) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Motch B; N2ECD; N2ICD; Neurogenic locus notch homolog protein 2; Neurogenic locus notch homolog protein 2 Notch 2 hN2 Notch 2 extracellular truncation Notch 2 intracellular domain NOTCH2; Notch 2; Notch 2 extracellular truncation; Notch 2 intracellular domain; Notch gene homolog 2; Notch homolog 2
Gene Aliases: AGS2; AI853703; HJCYS; hN2; N2; NOTCH2
UniProt ID: (Human) Q04721, (Mouse) O35516
Entrez Gene ID: (Human) 4853, (Mouse) 18129
Molecular Function: defense/immunity protein membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.