Abnova
Manufacturer Code:PAB10294
Catalog # 89-114-765
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 2396-2409 of human NOTCH2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of NOTCH2.
For Research Use Only
Protein Aliases: AGS2; hN2; N2ECD; N2ICD; Neurogenic locus notch homolog protein 2; Notch 2; Notch 2 extracellular truncation; Notch 2 intracellular domain; Notch homolog 2
Gene Aliases: AGS2; HJCYS; hN2; NOTCH2
UniProt ID: (Human) Q04721
Entrez Gene ID: (Human) 4853
Molecular Function: defense/immunity protein membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.