Invitrogen
Manufacturer Code:MA516862
Catalog # PIMA516862
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | mN1A |
Immunogen | Synthetic peptide corresponding to cdc10-NCR region within mouse Notch1. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4°C or -20°C if preferred |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: major type A protein; Motch A; mT14; Neurogenic locus notch homolog protein 1; NEXT; NICD; Notch 1; Notch 1 extracellular truncation; Notch 1 intracellular domain; Notch gene homolog 1; Notch homolog 1, translocation-associated; p300; RP23-306D20.12-001 9930111A19Rik Mis6 N1 Tan1 lin-12Motch A Notch gene homolog 1 mT14 major type A protein neurogenic locus notch homolog protein 1 p300 transmembrane receptor Notch1; Translocation-associated notch protein TAN-1; transmembrane receptor Notch1
Gene Aliases: 9930111A19Rik; AOS5; AOVD1; hN1; lin-12; Mis6; Motch; N1; NOTCH1; TAN1
UniProt ID: (Human) P46531, (Mouse) Q01705
Entrez Gene ID: (Human) 4851, (Mouse) 18128
Molecular Function:
defense/immunity protein
membrane-bound signaling molecule
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.