Abnova
Manufacturer Code:PAB17901
Catalog # 89-119-543
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the residues surrounding amino acid 602 of rat Nos3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Nos3.
For Research Use Only
Protein Aliases: cNOS; Constitutive NOS; EC-NOS; endothelial nitric oxide synthase 3; Endothelial NOS; eNOS; Nitric oxide synthase, endothelial; NOS type III; NOSIII
Gene Aliases: eNos; Nos3
UniProt ID: (Rat) Q62600
Entrez Gene ID: (Rat) 24600
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.