Novus Biologicals
Manufacturer Code:NBP273039AF405
Catalog # N273039A405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C6 |
Immunogen | Full length recombinant protein of human NORE1 (NP_872606) produced in HEK293T cells. |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Maxp1 MGC10823 MGC17344 New ras effector 1 NORE1A NORE1B NORE1RASSF3 Rap1-binding protein RAPLnovel Ras effector 1 Ras association (RalGDS/AF-6) domain family 5 Ras association (RalGDS/AF-6) domain family member 5 ras association domain-containing protein 5 Ras effector-like protein regulator for cell adhesion and polarization enriched in lymphoid tissue Regulator for cell adhesion and polarization enriched in lymphoid tissues tumor suppressor RASSF3; New ras effector 1; novel Ras effector 1; Rap1-binding protein; RAPL; Ras association (RalGDS/AF-6) domain family 5; Ras association (RalGDS/AF-6) domain family member 5; Ras association domain-containing protein 5; Ras effector-like protein; regulator for cell adhesion and polarization enriched in lymphoid tissue; Regulator for cell adhesion and polarization enriched in lymphoid tissues; tumor suppressor RASSF3
Gene Aliases: Maxp1; NORE1; NORE1A; NORE1B; RAPL; RASSF3; RASSF5
UniProt ID: (Human) Q8WWW0
Entrez Gene ID: (Human) 83593
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.