Invitrogen
Manufacturer Code:PA568825
Catalog # PIPA568825
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse NOR-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CHN CSMF MINOR NOR1 TEC chondrosarcoma extraskeletal myxoid fused to EWS mitogen-induced nuclear orphan receptor neuron-derived orphan receptor 1 nuclear hormone receptor NOR-1 nuclear receptor subfamily 4 group A member 3 translocated in extraskeletal chondrosarcoma; chondrosarcoma, extraskeletal myxoid, fused to EWS; Mitogen-induced nuclear orphan receptor; Neuron-derived orphan receptor 1; Neuron-derived orphan receptor 1/2; Nuclear hormone receptor NOR-1; Nuclear hormone receptor NOR-1/NOR-2; Nuclear receptor subfamily 4 group A member 3; nuclear receptor subfamily 4, group A, member 3; Orphan nuclear receptor TEC; Translocated in extraskeletal chondrosarcoma
Gene Aliases: AI573420; CHN; CSMF; MINOR; NOR-1; NOR-2; NOR1; NR4A3; TEC
UniProt ID: (Human) Q92570, (Rat) P51179, (Mouse) Q9QZB6
Entrez Gene ID: (Rabbit) 100344785, (Human) 8013, (Rat) 58853, (Mouse) 18124, (Guinea pig) 100715800
Molecular Function:
nuclear hormone receptor
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.